siRNA Details
| Gene Symbol: ZPLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175056.1 | NP_778226.1 | 3 | 397 | ||
| XM_017005703.1 | XP_016861192.1 | 4 | 579 | |||
| XM_017005704.1 | XP_016861193.1 | 3 | 435 | |||
| GenBank mRNA | AY090780.1 | - | 3 | 397 | ||
| BC031261.1 | - | 10 | 1131 | 5273136 | ||
| BC143817.1 | - | 4 | 493 | |||

ZPLD1
Genomic Map