siRNA Details
| Gene Symbol: WFIKKN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175575.5 | NP_783165.1 | 2 | 1962 | ||
| XM_017024172.1 | XP_016879661.1 | 2 | 1441 | |||
| GenBank mRNA | AF468657.1 | - | 2 | 1284 | ||
| AK127743.1 | - | 2 | 1622 | |||
| AK131196.1 | - | 532 | ||||
| AK298172.1 | - | 2 | 1441 | |||
| AK312821.1 | - | 2 | 1456 | |||
| AY358142.1 | - | 2 | 1797 | |||
| BC140842.1 | - | 2 | 1356 | 9021710 | ||
| BC140844.1 | - | 2 | 1356 | 9021712 | ||

WFIKKN2
Genomic Map