siRNA Details
| Gene Symbol: RABL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173825.3 | NP_776186.2 | 6, 7 | 644 | ||
| XM_017006227.1 | XP_016861716.1 | 6, 7 | 628 | |||
| GenBank mRNA | AK025772.1 | - | 3, 4 | 340 | ||
| AK293085.1 | - | 6, 7 | 628 | |||
| BC020832.1 | - | 6, 7 | 623 | 4771781 | ||
| BX647559.1 | - | 6, 7 | 641 | |||
| BX647593.1 | - | 6, 7 | 644 | |||
| BX647664.1 | - | 6, 7 | 628 | |||

RABL3
Genomic Map