siRNA Details
| Gene Symbol: RELL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130029.1 | NP_001123501.1 | 3 | 1077 | ||
| NM_173828.4 | NP_776189.3 | 2 | 1505 | |||
| XM_005268414.3 | XP_005268471.1 | 3 | 1081 | |||
| XM_011537624.1 | XP_011535926.1 | 3 | 1097 | |||
| XM_011537625.2 | XP_011535927.1 | 2 | 1509 | |||
| GenBank mRNA | AK054889.1 | - | 2 | 1505 | ||
| AK075064.1 | - | 2 | 332 | |||
| AY358163.1 | - | 2 | 1441 | |||
| BC063469.1 | - | 3 | 546 | 5211730 | ||

RELL2
Genomic Map