siRNA Details
| Gene Symbol: GPR32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001506.2 | NP_001497.1 | 1 | 1171 | ||
| GenBank mRNA | BC067453.1 | - | 1 | 1034 | 6971741 | |
| BC067454.1 | - | 1 | 1034 | 6971744 | ||
| BC095544.1 | - | 1 | 1172 | 6971954 | ||
More Information
Set Membership: |
|

GPR32
Genomic Map