siRNA Details
| Gene Symbol: GAS2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303130.1 | NP_001290059.1 | 5, 6 | 602 | ||
| NM_001303131.1 | NP_001290060.1 | 6, 7 | 1089 | |||
| NM_174942.2 | NP_777602.1 | 6, 7 | 723 | |||
| XM_005268804.3 | XP_005268861.1 | 7, 8 | 792 | |||
| XM_011538219.2 | XP_011536521.1 | 6, 7 | 648 | |||
| XM_011538220.2 | XP_011536522.1 | 5, 6 | 1983 | |||
| XM_011538221.2 | XP_011536523.1 | 5, 6 | 833 | |||
| XM_017019206.1 | XP_016874695.1 | 7, 8 | 866 | |||
| GenBank mRNA | AK095594.1 | - | 6, 7 | 1089 | ||
| AK315189.1 | - | 5, 6 | 580 | |||
| BC043366.1 | - | 6, 7 | 673 | 5104644 | ||

GAS2L3
Genomic Map