siRNA Details
| Gene Symbol: ZPLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175056.1 | NP_778226.1 | 11 | 2198 | ||
| XM_017005703.1 | XP_016861192.1 | 12 | 2380 | |||
| XM_017005704.1 | XP_016861193.1 | 11 | 2236 | |||
| GenBank mRNA | AY090780.1 | - | 11 | 2198 | ||
| BC021279.2 | - | 5 | 1330 | |||

ZPLD1
Genomic Map