siRNA Details
| Gene Symbol: AIRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000383.3 | NP_000374.1 | 7, 8 | 1056 | ||
| GenBank mRNA | AB006682.1 | - | 7, 8 | 1014 | ||
| AB006683.1 | - | 2 | 532 | |||
| AB006685.1 | - | 2 | 532 | |||
| BC137268.1 | - | 1 | 713 | 9021265 | ||
| BC137270.1 | - | 1 | 712 | 9021267 | ||
| Z97990.1 | - | 7, 8 | 1007 | |||

AIRE
Genomic Map