siRNA Details
Gene Symbol: TBXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001060.5 | NP_001051.1 | 3 | 1256 | ||
NM_201636.2 | NP_963998.2 | 3 | 1256 | |||
XM_011528214.2 | XP_011526516.1 | 4 | 3290 | |||
GenBank mRNA | AY429110.1 | - | 2 | 869 | ||
BC074749.2 | - | 2 | 908 | 30915459 | ||
BC074750.2 | - | 2 | 908 | 30915209 | ||
BC121148.1 | - | 2 | 514 | 40121265 | ||
U11271.1 | - | 3 | 1081 | |||
U27325.1 | - | 3 | 992 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
