siRNA Details
| Gene Symbol: KCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002237.3 | NP_002228.2 | 2 | 816 | ||
| XM_006723785.2 | XP_006723848.1 | 4 | 1199 | |||
| XM_011528800.1 | XP_011527102.1 | 3 | 874 | |||
| XM_011528801.1 | XP_011527103.1 | 4 | 983 | |||
| XM_011528802.1 | XP_011527104.1 | 4 | 978 | |||
| XM_011528803.2 | XP_011527105.1 | 4 | 905 | |||
| XM_011528804.1 | XP_011527106.1 | 3 | 1128 | |||
| XM_011528805.2 | XP_011527107.1 | 2 | 3039 | |||
| XM_011528806.1 | XP_011527108.1 | 2 | 1020 | |||
| GenBank mRNA | AF033383.1 | - | 1 | 521 | ||
| AK290689.1 | - | 2 | 817 | |||
| BC006367.1 | - | 3 | 1032 | 4131541 | ||
| BC046629.2 | - | 2 | 785 | 5287759 | ||
| CN301212.1 | - | 1 | 537 | |||

KCNG1
Genomic Map