siRNA Details
| Gene Symbol: APOE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000041.3 | NP_000032.1 | 4 | 385 | ||
| NM_001302688.1 | NP_001289617.1 | 4 | 467 | |||
| NM_001302689.1 | NP_001289618.1 | 4 | 316 | |||
| NM_001302690.1 | NP_001289619.1 | 4 | 416 | |||
| NM_001302691.1 | NP_001289620.1 | 4 | 400 | |||
| GenBank mRNA | AK314898.1 | - | 4 | 338 | ||
| BC003557.1 | - | 4 | 314 | 3355712 | ||
| BI550631.1 | - | 4 | 362 | |||
| BM042032.1 | - | 4 | 400 | |||
| BU848796.1 | - | 352 | ||||
| CA771535.1 | - | 3 | 534 | |||
| HY056112.1 | - | 4 | 385 | |||
| HY074384.1 | - | 4 | 316 | |||
| HY074888.1 | - | 4 | 416 | |||
| K00396.1 | - | 4 | 329 | |||
| KU177910.1 | - | 3 | 269 | |||
| KU177911.1 | - | 3 | 269 | |||
| M12529.1 | - | 4 | 330 | |||
| X00199.1 | - | 2 | 80 | |||

APOE
Genomic Map