siRNA Details
| Gene Symbol: SAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000331.5 | NP_000322.2 | 4 | 474 | ||
| NM_001178006.2 | NP_001171477.1 | 5 | 388 | |||
| NM_199161.4 | NP_954630.1 | 4 | 327 | |||
| GenBank mRNA | BC007022.1 | - | 4 | 288 | 4071612 | |
| BC105796.1 | - | 5 | 353 | 4722684 | ||
| BG533276.1 | - | 436 | ||||
| BG562565.1 | - | 4 | 327 | |||
| BG565078.1 | - | 4 | 436 | |||
| BG567902.1 | - | 379 | ||||
| BQ691948.1 | - | 3 | 190 | |||
| BX102362.1 | - | 4 | 394 | |||
| CD102084.1 | - | 4 | 291 | |||
| CR542241.1 | - | 3 | 251 | |||
| M10906.1 | - | 3 | 251 | |||
| M23698.1 | - | 3 | 251 | |||
| X51439.1 | - | 1 | 37 | |||
| X51441.1 | - | 1 | 37 | |||
| Gene Symbol: SAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X51442.1 | - | 1 | 37 | ||

SAA1
Genomic Map