siRNA Details
| Gene Symbol: GSTM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000848.3 | NP_000839.1 | 5 | 394 | ||
| NM_001142368.1 | NP_001135840.1 | 5 | 394 | |||
| GenBank mRNA | AK299482.1 | - | 5 | 523 | ||
| AK309796.1 | - | 5 | 311 | |||
| BC105038.1 | - | 5 | 325 | 8144041 | ||
| BC105066.1 | - | 5 | 325 | 8144069 | ||
| BC110380.1 | - | 5 | 347 | 5760414 | ||
| BM924181.1 | - | 5 | 360 | |||
| BT019947.1 | - | 5 | 300 | |||
| DA528798.1 | - | 5 | 394 | |||
| M63509.1 | - | 5 | 316 | |||

GSTM2
Genomic Map