siRNA Details
| Gene Symbol: ELANE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001972.3 | NP_001963.1 | 5 | 979 | ||
| XM_011527775.1 | XP_011526077.1 | 6 | 1656 | |||
| XM_011527776.1 | XP_011526078.1 | 6 | 995 | |||
| GenBank mRNA | BI254663.1 | - | 1 | 130 | ||
| D00187.1 | - | 4 | 769 | |||
| J03545.1 | - | 3 | 637 | |||
| M27783.1 | - | 2 | 493 | |||
| M34379.1 | - | 5 | 897 | |||
| X05875.1 | - | 5 | 897 | |||

ELANE
Genomic Map