siRNA Details
| Gene Symbol: FGF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002007.2 | NP_001998.1 | 3 | 801 | ||
| XM_005273847.1 | XP_005273904.1 | 2 | 458 | |||
| GenBank mRNA | AW340506.1 | - | 2 | 133 | ||
| FJ456981.1 | - | 2 | 457 | |||
| M17446.1 | - | 3 | 801 | |||

FGF4
Genomic Map