siRNA Details
| Gene Symbol: CNR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001841.2 | NP_001832.1 | 2 | 1222 | ||
| XM_011540629.2 | XP_011538931.1 | 2 | 1505 | |||
| XM_017000261.1 | XP_016855750.1 | 5 | 2197 | |||
| GenBank mRNA | AJ430063.1 | - | 1 | 1131 | ||
| BC069722.1 | - | 2 | 1164 | 7262528 | ||
| BC074767.2 | - | 1 | 1123 | 30915186 | ||
| EU517121.1 | - | 3 | 1354 | |||
| X74328.1 | - | 2 | 1221 | |||
More Information
Set Membership: |
|

CNR2
Genomic Map