siRNA Details
| Gene Symbol: PRB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261399.2 | NP_001248328.1 | 2 | 120 | ||
| NM_002723.5 | NP_002714.2 | 2 | 120 | |||
| GenBank mRNA | BC035362.1 | - | 2 | 106 | ||
| BC128191.1 | - | 2 | 120 | |||
| BC130386.1 | - | 2 | 105 | 40146417 | ||
| K03207.1 | - | 2 | 119 | |||

PRB4
Genomic Map