siRNA Details
| Gene Symbol: GP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000174.4 | NP_000165.1 | 3 | 879 | ||
| XM_005247374.3 | XP_005247431.1 | 5 | 1175 | |||
| XM_011512701.1 | XP_011511003.1 | 6 | 1257 | |||
| XM_011512702.1 | XP_011511004.1 | 5 | 1132 | |||
| GenBank mRNA | BC030229.1 | - | 3 | 844 | 5226377 | |
| X52997.1 | - | 3 | 876 | |||

GP9
Genomic Map