siRNA Details
| Gene Symbol: AIRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000383.3 | NP_000374.1 | 12, 13 | 1680 | ||
| GenBank mRNA | AB006682.1 | - | 12, 13 | 1638 | ||
| AB006683.1 | - | 6, 7 | 1156 | |||
| AB006685.1 | - | 7, 8 | 1074 | |||
| AK309572.1 | - | 12, 13 | 2271 | |||
| BC137268.1 | - | 5, 6 | 1337 | 9021265 | ||
| BC137270.1 | - | 5, 6 | 1336 | 9021267 | ||
| Z97990.1 | - | 12, 13 | 1631 | |||

AIRE
Genomic Map