siRNA Details
| Gene Symbol: PRB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002723.5 | NP_002714.2 | 3 | 553 | ||
| GenBank mRNA | BC035362.1 | - | 3 | 602 | ||
| BC130386.1 | - | 3 | 601 | 40146417 | ||
| K03207.1 | - | 3 | 552 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRB4| Gene Symbol: PRB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002723.5 | NP_002714.2 | 3 | 553 | ||
| GenBank mRNA | BC035362.1 | - | 3 | 602 | ||
| BC130386.1 | - | 3 | 601 | 40146417 | ||
| K03207.1 | - | 3 | 552 | |||