siRNA Details
| Gene Symbol: PSMB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002801.3 | NP_002792.1 | 3 | 349 | ||
| GenBank mRNA | AK298012.1 | - | 2 | 546 | ||
| AK312208.1 | - | 3 | 309 | |||
| BC017198.1 | - | 3 | 300 | 3143189 | ||
| BC052369.1 | - | 3 | 287 | 4054165 | ||
| BM845639.1 | - | 3 | 322 | |||
| BP289065.1 | - | 3 | 349 | |||
| BT019723.1 | - | 3 | 221 | |||
| BT019724.1 | - | 3 | 221 | |||
| CR456982.1 | - | 3 | 221 | |||
| Y13640.1 | - | 3 | 309 | |||

PSMB10
Genomic Map