siRNA Details
| Gene Symbol: RANBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278639.1 | NP_001265568.1 | 5, 6 | 976 | ||
| NM_002882.3 | NP_002873.1 | 5, 6 | 724 | |||
| XM_011530291.2 | XP_011528593.1 | 5, 6 | 1038 | |||
| GenBank mRNA | CA489474.1 | - | 5, 6 | 666 | ||
| CR456556.1 | - | 5, 6 | 552 | |||
| X83617.1 | - | 5, 6 | 600 | |||

RANBP1
Genomic Map