siRNA Details
| Gene Symbol: SLC30A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003459.4 | NP_003450.2 | 1, 2 | 274 | ||
| XM_005264547.1 | XP_005264604.1 | 1, 2 | 270 | |||
| XM_011533103.2 | XP_011531405.1 | 1, 2 | 262 | |||
| GenBank mRNA | AK313600.1 | - | 1, 2 | 229 | ||
| AL043136.2 | - | 1, 2 | 274 | |||
| BC028358.2 | - | 1, 2 | 262 | 4837595 | ||
| U76010.1 | - | 1, 2 | 171 | |||

SLC30A3
Genomic Map