siRNA Details
| Gene Symbol: NDUFB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004146.5 | NP_004137.2 | 2 | 289 | ||
| XM_011528039.2 | XP_011526341.1 | 2 | 881 | |||
| GenBank mRNA | AF112200.1 | - | 2 | 275 | ||
| AF217091.1 | - | 2 | 234 | |||
| BC002595.2 | - | 2 | 274 | 3140806 | ||
| BU565399.1 | - | 290 | ||||
| CB124371.1 | - | 2 | 289 | |||
| CR450354.1 | - | 2 | 212 | |||
| M33374.1 | - | 2 | 244 | |||

NDUFB7
Genomic Map