siRNA Details
| Gene Symbol: NDUFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004545.3 | NP_004536.2 | 2 | 267 | ||
| GenBank mRNA | AF050638.1 | - | 2 | 121 | ||
| AK055348.1 | - | 2 | 2193 | |||
| AK316557.2 | - | 2 | 889 | |||
| BC009691.1 | - | 2 | 127 | |||
| BC104672.1 | - | 2 | 187 | |||
| BC126232.1 | - | 2 | 264 | 8991948 | ||
| BC126234.1 | - | 2 | 264 | 8991950 | ||
| BM052981.1 | - | 2 | 267 | |||
| BX476462.1 | - | 2 | 166 | |||

NDUFB1
Genomic Map