siRNA Details
| Gene Symbol: HIST1H4I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003495.2 | NP_003486.1 | 1 | 334 | ||
| GenBank mRNA | BC016336.1 | - | 1 | 365 | ||
| BC066250.1 | - | 1 | 334 | 7002105 | ||
| BC067496.1 | - | 1 | 334 | 7002103 | ||
| BC067497.1 | - | 1 | 334 | 7002106 | ||
| BC075806.1 | - | 1 | 346 | |||

HIST1H4I
Genomic Map