siRNA Details
| Gene Symbol: HIST1H3J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003535.2 | NP_003526.1 | 1 | 356 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H3J| Gene Symbol: HIST1H3J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003535.2 | NP_003526.1 | 1 | 356 | ||