siRNA Details
| Gene Symbol: GFI1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004188.6 | NP_004179.3 | 8, 9 | 1334 | ||
| XM_006717297.3 | XP_006717360.1 | 4, 5 | 825 | |||
| XM_011519068.2 | XP_011517370.1 | 6, 7 | 1225 | |||
| XM_011519069.2 | XP_011517371.1 | 5, 6 | 995 | |||
| XM_011519070.2 | XP_011517372.1 | 6, 7 | 1225 | |||
| XM_017015175.1 | XP_016870664.1 | 5, 6 | 955 | |||
| GenBank mRNA | AF081946.1 | - | 3, 4 | 509 | ||
| AY428733.1 | - | 4, 5 | 660 | |||
| BC043371.1 | - | 8, 9 | 1334 | 5170920 | ||
| BC048254.1 | - | 3, 4 | 540 | |||
| CR536546.1 | - | 3, 4 | 509 | |||

GFI1B
Genomic Map