siRNA Details
| Gene Symbol: HMGCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098272.2 | NP_001091742.1 | 3 | 598 | ||
| NM_001324219.1 | NP_001311148.1 | 2 | 585 | |||
| NM_001324220.1 | NP_001311149.1 | 3 | 618 | |||
| NM_001324222.1 | NP_001311151.1 | 3 | 598 | |||
| NM_001324223.1 | NP_001311152.1 | 2 | 539 | |||
| NM_001324224.1 | NP_001311153.1 | 3 | 618 | |||
| NM_002130.7 | NP_002121.4 | 2 | 539 | |||
| XM_017009412.1 | XP_016864901.1 | 3 | 678 | |||
| GenBank mRNA | AK095492.1 | - | 2, 3 | 439 | ||
| AK315593.1 | - | 3 | 526 | |||
| BC000297.2 | - | 3 | 477 | |||
| BC083514.1 | - | 2 | 437 | 4443624 | ||
| BJ995153.1 | - | 3 | 582 | |||
| BT007302.1 | - | 1 | 372 | |||
| DB074885.1 | - | 2 | 540 | |||
| DB078662.1 | - | 2 | 520 | |||
| L25798.1 | - | 1 | 393 | |||
| X66435.1 | - | 3 | 494 | |||

HMGCS1
Genomic Map