siRNA Details
| Gene Symbol: FKBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135208.1 | NP_001128680.1 | 3 | 303 | ||
| NM_004470.3 | NP_004461.2 | 3 | 388 | |||
| NM_057092.2 | NP_476433.1 | 3 | 294 | |||
| XM_005273848.3 | XP_005273905.1 | 3 | 290 | |||
| GenBank mRNA | BC003384.1 | - | 3 | 294 | 3445148 | |
| BC091475.1 | - | 3 | 240 | 30349098 | ||
| BT009861.1 | - | 2 | 192 | |||
| CR407636.1 | - | 2 | 192 | |||
| M65128.1 | - | 3 | 226 | |||
| M75099.1 | - | 3 | 222 | |||

FKBP2
Genomic Map