siRNA Details
| Gene Symbol: CLIC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004669.2 | NP_004660.2 | 4 | 370 | ||
| XM_017015281.1 | XP_016870770.1 | 4 | 575 | |||
| XM_017015282.1 | XP_016870771.1 | 4 | 538 | |||
| GenBank mRNA | AF102166.1 | - | 4 | 330 | ||
| BC007012.1 | - | 4 | 366 | 3996920 | ||
| CR542066.1 | - | 3 | 254 | |||

CLIC3
Genomic Map