siRNA Details
Gene Symbol: HOXA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002141.4 | NP_002132.3 | 2 | 938 | ||
GenBank mRNA | BC126994.1 | - | 2 | 338 | ||
M74297.1 | - | 2 | 891 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HOXA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002141.4 | NP_002132.3 | 2 | 938 | ||
GenBank mRNA | BC126994.1 | - | 2 | 338 | ||
M74297.1 | - | 2 | 891 |