siRNA Details
| Gene Symbol: SEPW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003009.2 | NP_003000.1 | 3 | 281 | ||
| GenBank mRNA | AF015283.1 | - | 3 | 130 | ||
| AK026717.1 | - | 2 | 1843 | |||
| BC000581.2 | - | 3 | 146 | 3162433 | ||
| BC032546.1 | - | 3 | 171 | 5535970 | ||
| BC039597.1 | - | 3 | 149 | 6015153 | ||
| BC047893.1 | - | 3 | 148 | 6204763 | ||
| BF973456.1 | - | 3 | 294 | |||
| U67171.1 | - | 3 | 141 | |||

SEPW1
Genomic Map