siRNA Details
| Gene Symbol: IFITM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003641.3 | NP_003632.3 | 2 | 695 | ||
| GenBank mRNA | BC000897.1 | - | 2 | 645 | 3464598 | |
| J04164.1 | - | 2 | 831 | |||
| X84958.1 | - | 2 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFITM1| Gene Symbol: IFITM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003641.3 | NP_003632.3 | 2 | 695 | ||
| GenBank mRNA | BC000897.1 | - | 2 | 645 | 3464598 | |
| J04164.1 | - | 2 | 831 | |||
| X84958.1 | - | 2 | 625 | |||