siRNA Details
| Gene Symbol: GFI1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135031.1 | NP_001128503.1 | 5 | 798 | ||
| NM_004188.6 | NP_004179.3 | 10 | 1610 | |||
| XM_006717297.3 | XP_006717360.1 | 6 | 1167 | |||
| XM_011519068.2 | XP_011517370.1 | 8 | 1567 | |||
| XM_011519069.2 | XP_011517371.1 | 7 | 1337 | |||
| XM_011519070.2 | XP_011517372.1 | 8 | 1501 | |||
| XM_017015175.1 | XP_016870664.1 | 7 | 1297 | |||
| XM_017015176.1 | XP_016870665.1 | 7 | 1363 | |||
| GenBank mRNA | AF081946.1 | - | 5 | 785 | ||
| AY428733.1 | - | 6 | 936 | |||
| BC043371.1 | - | 10 | 1610 | 5170920 | ||
| BC048254.1 | - | 5 | 816 | |||
| CR536546.1 | - | 5 | 785 | |||

GFI1B
Genomic Map