siRNA Details
| Gene Symbol: CLIC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004669.2 | NP_004660.2 | 6 | 670 | ||
| XM_017015281.1 | XP_016870770.1 | 6 | 875 | |||
| XM_017015282.1 | XP_016870771.1 | 6 | 838 | |||
| GenBank mRNA | AF102166.1 | - | 6 | 630 | ||
| BC007012.1 | - | 6 | 666 | 3996920 | ||
| CR542066.1 | - | 5 | 554 | |||

CLIC3
Genomic Map