siRNA Details
| Gene Symbol: CRYGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005210.3 | NP_005201.2 | 3 | 552 | ||
| XM_017003402.1 | XP_016858891.1 | 3 | 732 | |||
| GenBank mRNA | BC074944.2 | - | 3 | 552 | 30915273 | |
| BC074945.2 | - | 3 | 552 | 30915496 | ||
| BC117384.1 | - | 3 | 552 | 40125935 | ||
| BC117388.1 | - | 3 | 552 | 40125939 | ||
| KT326704.1 | - | 3 | 557 | |||

CRYGB
Genomic Map