siRNA Details
| Gene Symbol: SNAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006049.2 | NP_006040.1 | 1, 2 | 175 | ||
| GenBank mRNA | AF093593.1 | - | 1, 2 | 98 | ||
| AK024763.1 | - | 1, 2 | 147 | |||
| AK312018.1 | - | 1, 2 | 147 | |||
| CD685245.1 | - | 1, 2 | 212 | |||
| CN347010.1 | - | 1, 2 | 97 | |||
| DB037177.1 | - | 1, 2 | 107 | |||

SNAPC5
Genomic Map