siRNA Details
| Gene Symbol: TTLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037162.1 | 11 | 2829 | |||
| GenBank mRNA | AK297289.1 | - | 10 | 1966 | ||
| AL096725.1 | - | 5 | 2154 | |||
| AL833939.1 | - | 8 | 2944 | |||
| BC098298.1 | - | 6 | 1764 | 40025661 | ||
| Gene Symbol: ARPC4-TTLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198793.1 | NP_001185722.1 | 12 | 2656 | ||
| GenBank mRNA | BC025289.1 | - | 11 | 2090 | ||

TTLL3
Genomic Map