siRNA Details
| Gene Symbol: DHRS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005794.3 | NP_005785.1 | 4, 5 | 860 | ||
| NM_182908.4 | NP_878912.1 | 4, 5 | 860 | |||
| XM_005267249.1 | XP_005267306.1 | 4, 5 | 572 | |||
| XM_006720001.3 | XP_006720064.1 | 4, 5 | 535 | |||
| XM_011536338.1 | XP_011534640.1 | 3, 4 | 501 | |||
| XM_011536339.2 | XP_011534641.1 | 3, 4 | 351 | |||
| GenBank mRNA | AK222857.1 | - | 4, 5 | 860 | ||
| BC002786.2 | - | 4, 5 | 851 | 3614599 | ||
| BC007339.2 | - | 3, 4 | 386 | |||
| U31875.1 | - | 4, 5 | 846 | |||

DHRS2
Genomic Map