siRNA Details
| Gene Symbol: HS3ST3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006041.2 | NP_006032.1 | 2 | 1776 | ||
| XM_017025479.1 | XP_016880968.1 | 2 | 2204 | |||
| GenBank mRNA | AF105377.1 | - | 2 | 1637 | ||
| BC063301.1 | - | 2 | 1693 | 30343352 | ||
| BC069664.1 | - | 2 | 1577 | 7262483 | ||
| BC069725.1 | - | 2 | 1576 | 7262495 | ||

HS3ST3B1
Genomic Map