siRNA Details
| Gene Symbol: DUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012146.1 | NP_036278.1 | 1 | 1141 | ||
| GenBank mRNA | AJ001481.1 | - | 1 | 1141 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUX1| Gene Symbol: DUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012146.1 | NP_036278.1 | 1 | 1141 | ||
| GenBank mRNA | AJ001481.1 | - | 1 | 1141 | ||