siRNA Details
| Gene Symbol: VPREB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013378.2 | NP_037510.1 | 2 | 413 | ||
| GenBank mRNA | AF163825.1 | - | 2 | 375 | ||
| AK312098.1 | - | 2 | 368 | |||
| AY359000.1 | - | 2 | 413 | |||
| BC020666.1 | - | 2 | 438 | 4772032 | ||
| BQ899182.1 | - | 2 | 346 | |||
| CR456610.1 | - | 2 | 362 | |||

VPREB3
Genomic Map