siRNA Details
| Gene Symbol: CLDN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185072.2 | NP_001172001.1 | 4 | 892 | ||
| NM_001185073.2 | NP_001172002.1 | 3 | 853 | |||
| NM_012129.4 | NP_036261.1 | 3 | 849 | |||
| GenBank mRNA | AK312480.1 | - | 4 | 777 | ||
| AL136770.1 | - | 3 | 701 | |||
| BC036754.2 | - | 2 | 638 | 5201423 | ||
| BC068532.1 | - | 3 | 735 | 5294796 | ||
| CR533470.1 | - | 1 | 510 | |||

CLDN12
Genomic Map