siRNA Details
Gene Symbol: ZNF117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015852.3 | NP_056936.2 | 4 | 2409 | ||
GenBank mRNA | BC034021.1 | - | 9 | 1562 | ||
M55422.1 | - | 4 | 2408 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015852.3 | NP_056936.2 | 4 | 2409 | ||
GenBank mRNA | BC034021.1 | - | 9 | 1562 | ||
M55422.1 | - | 4 | 2408 |