siRNA Details
| Gene Symbol: HES6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142853.2 | NP_001136325.1 | 4 | 1085 | ||
| NM_001282434.1 | NP_001269363.1 | 4 | 1051 | |||
| NM_018645.5 | NP_061115.2 | 4 | 1091 | |||
| GenBank mRNA | AF260237.1 | - | 4 | 932 | ||
| AK075040.1 | - | 3 | 1240 | |||
| BC007939.2 | - | 4 | 996 | 4131881 | ||

HES6
Genomic Map