siRNA Details
| Gene Symbol: HOXC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018953.3 | NP_061826.1 | 1 | 500 | ||
| GenBank mRNA | BM786543.1 | - | 1 | 413 | ||
| CN309383.1 | - | 1 | 76 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_175730, BC115551, AK078527, BC115552.

HOXC5
Genomic Map