siRNA Details
| Gene Symbol: OPN1LW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020061.5 | NP_064445.2 | 2 | 397 | ||
| GenBank mRNA | BQ637240.1 | - | 1 | 71 | ||
| BQ639996.1 | - | 2 | 340 | |||
| CR749814.1 | - | 2 | 355 | |||
| DN693335.1 | - | 2 | 397 | |||
| EL952084.1 | - | 2 | 372 | |||
More Information
Set Membership: |
|

OPN1LW
Genomic Map