siRNA Details
| Gene Symbol: CARD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021571.3 | NP_067546.1 | 1 | 109 | ||
| GenBank mRNA | AF208005.1 | - | 1 | 101 | ||
| AY358231.1 | - | 1 | 113 | |||
| BC131610.1 | - | 1 | 101 | 40115072 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CARD18| Gene Symbol: CARD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021571.3 | NP_067546.1 | 1 | 109 | ||
| GenBank mRNA | AF208005.1 | - | 1 | 101 | ||
| AY358231.1 | - | 1 | 113 | |||
| BC131610.1 | - | 1 | 101 | 40115072 | ||