siRNA Details
| Gene Symbol: GAL3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022134.2 | NP_071417.2 | 4 | 520 | ||
| GenBank mRNA | AB040610.1 | - | 4 | 521 | ||
| BC117293.1 | - | 4 | 438 | 40125844 | ||
| BC117295.1 | - | 4 | 438 | 40125846 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAL3ST2| Gene Symbol: GAL3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022134.2 | NP_071417.2 | 4 | 520 | ||
| GenBank mRNA | AB040610.1 | - | 4 | 521 | ||
| BC117293.1 | - | 4 | 438 | 40125844 | ||
| BC117295.1 | - | 4 | 438 | 40125846 | ||